missing translation for 'onlineSavingsMsg'
Learn More

Thrombomodulin/BDCA-3 Antibody (THBD/1782), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18402084 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402084 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402084 Supplier Novus Biologicals Supplier No. NBP254496JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombomodulin/BDCA-3
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Classification Monoclonal
Clone THBD/1782
Conjugate Janelia Fluor 549
Formulation 50 mM Sodium Borate
Gene Alias BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
Host Species Mouse
Immunogen Recombinant fragment (around aa 69-194) of human Thrombomodulin/BDCA-3 protein (exact sequence is proprietary) (Uniprot: P07204)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7056
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.