missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin/BDCA-3 Antibody (THBD/1591), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54490H
This item is not returnable.
View return policy
Description
Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifications
| Thrombomodulin/BDCA-3 | |
| Monoclonal | |
| HRP | |
| BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
| Recombinant fragment (around aa 69-194) of human Thrombomodulin/BDCA-3 protein (exact sequence is proprietary) (Uniprot: P07204) | |
| 100 μg | |
| Cardiovascular Biology | |
| 7056 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| THBD/1591 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction