missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thrombo-Wellcotest™ Rapid Latex Agglutination Test

Product Code. 13215669
Click to view available options
Quantity:
100 Tubes/Pk.
100 mL/Bottle
20 Tests/Kit
3 mL/Vial
Type:
Collection Tubes
Glycine Saline Buffer
Kit
Latex
Unit Size:
100mL
20 tests
3mL
Pack of 100
This item is not returnable. View return policy

Product Code. 13215669

Brand: Thermo Scientific™ R30852301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapidly detect fibrin and fibrinogen degradation products in human serum and urine by latex agglutination.

Carry out semi-quantitative detection of fibrin and fibrinogen degradation products in human serum and urine using Thermo Scientific™ Thrombo-Wellcotest™ Rapid Latex Agglutination Test.

Use this simple, rapid, semi-quantitative test for the presence of fibrin/fibrinogen degredation products (FDP) in human serum and urine samples.

The test is designed as a slide agglutination method in which one drop of sample and one drop of latex suspension are mixed. A distinct agglutination pattern at the end of the test period indicates the presence of 2μg/ml FDP or more in the sample under test. By assaying samples at two different dilutions, an approximate FDP value can be determined.

Capable of detecting the presence of all the major breakdown products of fibrin or fibrinogen. The levels measured by this latex test correlate well with results obtained with a haemagglutination-inhibition immunoassay.

The following products are available for use with Thrombo-Wellcotest Test Kit and are sold separately:

  • Thrombo-Wellcotest Latex (Part No. R30852301)
  • Thrombo-Wellcotest Collection Tubes (Part No. R30853001)—Glass tubes contain soya bean trypsin inhibitor (approximately 3600 NF units/tube) and Bothrops atrox venom (>10μg/tube) for the collection of 2mL whole blood or urine.
  • Thrombo-Wellcotest Glycine Saline Buffer (Part No. R30853101)—Glycine saline buffer is provided as a replacement item in instances where multiple tests have exhausted the supply of this component.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Format Vial
Type Latex
Quantity 3 mL/Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.