missing translation for 'onlineSavingsMsg'
Learn More

Threonine (L-Threonine), Mikromol™

Product Code. 30051441
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051441 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051441 Supplier Mikromol Supplier No. MM0360.000250
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Threonine (L-Threonine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Threonine
CAS 72-19-5
Analyte or Component Names L-Threonine
Shipping Condition Room Temperature
Molecular Formula C4 H9 N O3
Certifications/Compliance ISO 17034
Synonym Threonine, (2S,3R)-2-amino-3-hydroxybutanoic acid, (S)-Threonine, 17: PN: WO2011044089 SEQID: 18 claimed protein, 2-Amino-3-hydroxybutyric acid, 2: PN: US20090069547 PAGE: 10 claimed protein, 3046: PN: WO2004111636 SEQID: 516 unclaimed protein, 46: PN: WO2005016244 PAGE: 70 claimed protein, 48: PN: WO2004076659 FIGURE: 7 claimed protein, 49: PN: WO2005016244 PAGE: 70 claimed protein, 4: PN: WO2012076822 PAGE: 51 claimed sequence, Butanoic acid, 2-amino-3-hydroxy-, [R-(R*,S*)]-, L-(-)-Threonine, NSC 16589, NSC 46701, Threonin, [R-(R*,S*)]-2-Amino-3-hydroxybutanoic acid
InChI Formula InChI=1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m1/s1
SMILES [C@H]([C@@H](C)O)(C(O)=O)N
IUPAC Name (2S,3R)-2-amino-3-hydroxybutanoic acid
Molecular Weight (g/mol) 119.12
Quantity 250 mg
Formula Weight 119.0582 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.