missing translation for 'onlineSavingsMsg'
Learn More

threo-Dihydro Bupropion Hydrochloride, TRC

Product Code. 30384119
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384119 1 mg
30384580 10 mg
30387681 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384119 Supplier Toronto Research Chemicals Supplier No. TRCD4504731MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 292055-71-1
Molecular Formula C13 H20 Cl N O . HCl
Molecular Weight (g/mol) 278.21794
Synonym (alphaS)-3-Chloro-alpha-[(1S)-1-[(1,1-dimethylethyl)amino]ethyl]-benzenemethanol Hydrochloride, BW-A 494U, (R*,R*)-3-Chloro-alpha-[1-[(1,1-dimethylethyl)amino]ethyl]benzenemethanol Hydrochloride, (alphaR)-rel-3-Chloro-alpha-[(1R)-1-[(1,1-dimethylethyl)amino]ethyl]benezenemethanol Hydrochloride, (alphaS)-rel-3-chloro-alpha-[(1S)-1-[(1,1-dimethylethyl)amino]ethyl]benzenemethanol Hydrochloride
IUPAC Name (1S,2S)-2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-ol;hydrochloride
SMILES [C@H]([C@@H](NC(C)(C)C)C)(O)C1=CC(Cl)=CC=C1.Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C13H20ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,12,15-16H,1-4H3;1H/t9-,12+;/m0./s1
Formula Weight 277.1
Chemical Name or Material (S,S)-Dihydro Bupropion Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.