Learn More
Codan™ Three-part Syringes
Product Code. 15895777
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
Connection Type:
Centric Luer Cone
Luer Tip
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Connection Type unitSize
15895777 2 mL Centric Luer Cone Pack of 100
11338763 1 mL Luer Tip Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15895777 Supplier Codan™ Supplier No. 62.2611
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Codan™ Three-part Syringes to accommodate a variety of medical needs including: catheterization, hypodermic ear treatments, and more. The syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spill
  • Sterile
  • Graduations
  • Disposable
  • Latex-free
Specifications
Autoclavable Autoclavable
Material Polypropylene
Description Dual scale; Silicone seal
Product Type Hypodermic
Disposable Single-use
Graduations 0.1 mL
Volume (Metric) 2 mL
Connection Type Centric Luer Cone
Graduated Yes
Sterility Sterile