Learn More
Codan™ Three-part Syringes
Description
- Sterile
- Graduations
- Disposable
- Latex-free
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Description | Dual scale; Silicone seal |
| Product Type | Polypropylene |
| Disposable | Single-use |
| Volume (Metric) | 1 mL |
| Connection Type | Luer Tip |
| Graduated | Yes |
| Sterility | Sterile |