Learn More
SCAT Thread Adapter, PP
Product Code. 10363580 Shop All SCAT Europe Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Product Code. Material Unit Size
10363580 Polypropylene Each
1 options
Product Code. 10363580 Supplier SCAT Supplier No. 107095
  • ONLINE EXCLUSIVE PRICE
    £48.30
    £43.74 / Pack of 1
    Save £4.56
    9% Off
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
SCAT Safety Caps and SCAT Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
SCAT thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Suitable for all SCAT caps, funnels or canisters
  • Specifications
    Closure Size S51 (f) to S55 (m)
    Color Colorless
    Material Polypropylene
    Product Type Thread Adapter