Learn More
SymLine™ Thread Adapter
Description
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Specifications
| Closure Size | GL14, NPT 1/4 in. |
| Closure Color | Black |
| Liner Material | PTFE-EL |
| Product Type | Thread Adapter |