missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRAP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538545
This item is not returnable.
View return policy
Description
This gene encodes a component of the mediator complex , a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| THRAP4 | |
| Polyclonal | |
| Unconjugated | |
| MED24 | |
| THRAP4, MED24, thyroid hormone receptor associated protein 4, ARC100, DRIP100, KIAA0130, MGC8748, TRAP100, activator-recruited cofactor 100 kDa component, mediator of RNA polymerase II transcription, subunit 24 homolog, thyroid hormone receptor-associate | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23989, 619436, 9862 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75448, Q99K74, Q4V8B3 | |
| MED24 | |
| A synthetic peptide derived from the internal region of human MED24 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction