missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
THRAP2 Polyclonal specifically detects THRAP2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | THRAP2 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | KIAA1025thyroid hormone receptor associated protein 2, mediator complex subunit 13-likeThyroid hormone receptor-associated protein complex 240 kDa component-like, PROSIT240FLJ21627, THRAP2mediator of RNA polymerase II transcription subunit 13-like, Thyroid hormone receptor-associated protein 2, TRAP240LDKFZp781D0112 |
| Gene Symbols | MED13L |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PALYGNGLELQQLSTLDDRTVLVGQRLPLMAEVSETALYCGIRPSNPESSEKWWHSYRLPPSDDAEFRPPELQGERCDAKMEVNSESTALQRLLAQPNKRFKIWQD |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?