missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10139-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a nuclear hormone receptor for triiodothyronine. It is one of the several receptors for thyroid hormone, and has been shown to mediate the biological activities of thyroid hormone. Knockout studies in mice suggest that the different receptors, while having certain extent of redundancy, may mediate different functions of thyroid hormone. Alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
| THRA | |
| Polyclonal | |
| Unconjugated | |
| THRA | |
| AR7, c ERBA 1, c erbA alpha, EAR 7, EAR7, ERB T 1, ERBA, ERBA1, NR1A1, THRA, THRA1, THRA2, Thyroid hormone receptor alpha, V erbA related protein 7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21833, 7067, 81812 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.81 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P10827, P63058, P63059 | |
| THRA | |
| THRA Fusion Protein Ag0182 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction