missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ THPTA
GREENER_CHOICE

Product Code. 18706225
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18706225

Brand: Tocris Bioscience™ 7863/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Click chemistry auxiliary reagent; accelerating ligand for copper-catalyzed azide-alkyne cycloaddition reaction

THPTA is a click chemistry auxiliary reagent. It is a stabilizing ligand for copper-catalyzed azide-alkyne cycloaddition reactions.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris[(1-(3-hydroxypropyl)-1 H-1,2,3-triazol-4-yl]methyl)amine
CAS 760952-88-3
Quantity 100 mg
Target General Laboratory Research Reagent
Molecular Formula C18H30N10O3
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.