missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ThPOK Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA529461

Product Code. 13483447

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: 293T, HeLa, HepG2. Predicted reactivity: Mouse (87%), Rat (87%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZFP67 is an early growth response gene that encodes a zinc finger-containing transcription factor that binds to the promoter regions of type I collagen genes (e.g., COL1A1; MIM 120150) and has a role in development.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ThPOK
Polyclonal
Unconjugated
Zbtb7b
CKROX; c-KROX; DKFZp686G01254; hcKROX; Krueppel-related zinc finger protein cKrox; RP11-307C12.9; T helper-inducing POZ/Krueppel factor; T-helper-inducing POZ/Krueppel-like factor; Thpok; Vzinc finger and BTB domain containing 7B; ZBTB15; ZBTB7B; ZFP67; Zfp-67; zinc finger and BTB domain containing 15; zinc finger and BTB domain containing 7B; Zinc finger and BTB domain-containing protein 15; Zinc finger and BTB domain-containing protein 7B; zinc finger protein 67; Zinc finger protein 67 homolog; zinc finger protein 857B; Zinc finger protein Th-POK; ZNF857B
Rabbit
Antigen affinity chromatography
RUO
51043
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
O15156
Zbtb7b
Recombinant protein encompassing a sequence within the center region of human ThPok. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.