missing translation for 'onlineSavingsMsg'
Learn More

Thorium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™

Product Code. 11436008 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
This item is not returnable. View return policy

Product Code. 11436008

Brand: SPEX Certiprep CLTH22Y

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Th1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7440-29-1, 7697-37-2, 7732-18-5
Certificate of Analysis Claritas PPT Certificate of Analysis reports up to 68 elements at ppt levels.
CAS Min % 0.1, 2.0, 97.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.1, 2.0, 97.9
Molecular Formula Th
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key ZSLUVFAKFWKJRC-UHFFFAOYSA-N
SMILES [Th]
IUPAC Name thorium
Solvent or Matrix 2% HNO3
Molecular Weight (g/mol) 232.038
PubChem CID 23960
Quantity 125 mL
ChEBI CHEBI:33385
Grade Trace ICP/ICP-MS
Chemical Name or Material Claritas PPT Grade (Depleted) Thorium Standard
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : nitric acid, Thorium

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.