missing translation for 'onlineSavingsMsg'
Learn More

Thomsen-Friedenreich Antigen Antibody (A68-B/A11) - Azide and BSA Free, Novus Biologicals™

Product Code. 18367573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367573 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367573 Supplier Novus Biologicals Supplier No. NBP311557

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Thomsen-Friedenreich Antigen Monoclonal antibody specifically detects Thomsen-Friedenreich Antigen in Human, Mouse, Rat samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thomsen-Friedenreich Antigen
Applications ELISA
Classification Monoclonal
Clone A68-B/A11
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA 1-5 ug/ml
Formulation 10 mM PBS
Host Species Mouse
Immunogen Neuraminidase-treated human red blood cells
Purification Method Bioreactor concentrate
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.