missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THOC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545977
This item is not returnable.
View return policy
Description
Peptide sequence: ELEHLSHIKE SVEDKLELRR KQFHVLLSTI HELQQTLEND EKLSEVEEAQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 100%; Zebrafish: 90%.
THOC7 gene ontology annotations related to this gene include mRNA processing.
Specifications
| THOC7 | |
| Polyclonal | |
| Unconjugated | |
| THOC7 | |
| FLJ23445; fSAP24; Functional spliceosome-associated protein 24; hTREX30; hypothetical protein LOC445035; im:7153996; Ngg1 interacting factor 3 like 1 binding protein 1; ngg1-interacting factor 3-like protein 1-binding protein 1; NIF3L1-binding protein 1; Nif3l1bp1; RGD1305475; THO complex 7; THO complex 7 homolog; THO complex 7 homolog (Drosophila); THO complex subunit 7 homolog; THO complex subunit 7-like protein; THOC7; THOC7 protein; wu:fa20e08; wu:fj35e07; zgc:92711 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80145 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6I9Y2 | |
| THOC7 | |
| Synthetic peptide directed towards the C-terminal of human THOC7 (aa 145-194). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction