Learn More
Invitrogen™ THOC6 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA598560
Product Code. 16571382
  • £421.00 / 100µL
Description
THOC6 Polyclonal Antibody for Western Blot, ELISA
THOC6 belongs to the WD repeat THOC6 family. It contains 7 WD repeats. The function of the THOC6 protein remains unknown.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
THOC6
Polyclonal
Unconjugated
THOC6
BBIS; F830014G06Rik; fSAP35; Functional spliceosome-associated protein 35; hypothetical protein LOC450075; Pdrp; PSEC0006; THO complex 6; THO complex 6 homolog; THO complex 6 homolog (Drosophila); THO complex subunit 6 homolog; thoc6; WD repeat domain 58; WD repeat-containing protein 58; wdr58; zgc:101618
Rabbit
Protein G
RUO
79228
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot
3.42 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q86W42
THOC6
Recombinant Human THO complex subunit 6 homolog protein (56-315AA).