missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Immunology, Innate Immunity |
| Target | THOC5 |
| Synonym | C22orf19, chromosome 22 open reading frame 19, Fmip, Fms-interacting protein, fSAP79hTREX90, Functional spliceosome-associated protein 79, KIAA0983, NF2/meningioma region protein pK1.3, PK1.3, Placental protein 39.2, PP39.2, THO complex 5, THO complex subunit 5 homolog |
| Gene Symbol | THOC5 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?