missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THOC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111210
This item is not returnable.
View return policy
Description
Immunogen sequence: KTHRSKAEEQ FKFEVNEISW NNDNNMFFLT NGNGCINILS YPELKPVQSI NAHPSNCICI KFDPMGKYFA TGSADALV Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
THOC3 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.TEX1 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.
Specifications
| THOC3 | |
| Polyclonal | |
| Unconjugated | |
| THOC3 | |
| 2410044K02Rik; AL033344; hTREX45; LOC529231 protein; TEX1; TEX1 homolog; THO complex 3; THO complex subunit 3; Tho3; Thoc3; zgc:100815 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84321 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96J01 | |
| THOC3 | |
| Recombinant Protein corresonding to Human THOC3. Recombinant protein control fragment (Product #RP-99036). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction