missing translation for 'onlineSavingsMsg'
Learn More

THOC3 Antibody (OTI4H6) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30688611 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688611 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688611 Supplier Novus Biologicals Supplier No. NBP344944JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

THOC3 Monoclonal antibody specifically detects THOC3 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen THOC3
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI4H6
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias hTREX45, MGC5469, TEX1 homolog, TEX1THO complex subunit 3, THO complex 3, tho3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 15-237 of human THOC3(NP_115737) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 84321
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.