missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
THOC3 Monoclonal antibody specifically detects THOC3 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | THOC3 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI4H6 |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | hTREX45, MGC5469, TEX1 homolog, TEX1THO complex subunit 3, THO complex 3, tho3 |
| Host Species | Mouse |
| Immunogen | Human recombinant protein fragment corresponding to amino acids 15-237 of human THOC3(NP_115737) produced in E.coli. |
| Purification Method | >95% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?