missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ THOC3 293T Cell Transient Overexpression Lysate (Denatured)
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | THOC3 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | NM_032361 |
| Molecular Weight (g/mol) | Theoretical M.W. 38.8kDa |
| Plasmid | pCMV-THOC3 full-length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-THOC3 antibody (Mfr. No. H00084321-B02) by Western Blot. |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Host Species | Human |
| Show More |
Safety and Handling
Product Identifier
- 100UL THOC3 293T Cell Transient Overexpression Lys
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Mercaptoethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu