Learn More
Invitrogen™ THOC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110485
Description
Immunogen sequence: MAAAAVVVPA EWIKNWEKSG RGEFLHLCRI LSENKSHDSS TYRDFQQALY ELSYHVIKGN LKHEQASNVL SDISEFREDM PSILADVFCI LDIETNCLEE KSKRDYFTQL VLACLYLVSD TVLKERLDPE TLESLGLIKQ SQQFNQKSVK IKTKLFYKQQ KFNLLREENE GYAKLIAELG QDLSGSITSD LILENIKSLI GCFNLDPNRV LDVILEVFEC RPEHDDFFIS LLESYMSMCE PQTLCHILGF KFKFYQEPNG ETPSSLYRVA.
The TREX multiprotein complex binds specifically to spliced mRNAs to facilitate mRNA export. The protein encoded by this gene is a member of the THO complex, a subset of the TREX complex. The encoded protein interacts with the THOC1 protein.
Specifications
| THOC2 | |
| Polyclonal | |
| Unconjugated | |
| THOC2 | |
| 6330441O12Rik; CXorf3; D130005M13Rik; dJ506G2.1; Gm1139; Gm1793; hTREX120; mental retardation, X-linked 12; mental retardation, X-linked 35; MRX12; MRX35; RGD1561623; THO complex 2; THO complex subunit 2; Tho2; THOC2; transcription/export complex subunit 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 331401, 57187 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| B1AZI6, Q8NI27 | |
| THOC2 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.