missing translation for 'onlineSavingsMsg'
Learn More

THK5351 (R enantiomer), MedChemExpress

Product Code. 16436017
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16436017 5 mg 5mg
16420739 1 mg 1mg
16403298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16436017 Supplier Medchem Express Supplier No. HY101183A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress THK5351 R enantiomer is an R enantiomer of THK5351.

THK5351 R enantiomer is an R enantiomer of THK5351.

Specifications

Chemical Name or Material THK5351 (R enantiomer)
CAS 2101218-44-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H18FN3O2
Quantity 5 mg
SMILES CNC1=NC=C(C2=NC3=CC=C(OC[C@H](CF)O)C=C3C=C2)C=C1
Molecular Weight (g/mol) 327.35
Formula Weight 327.35
Percent Purity 95.62%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.