missing translation for 'onlineSavingsMsg'
Learn More

Thiosultap Sodium Salt, TRC

Product Code. 30455709
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455709 500 mg 500mg
30453155 100 mg 100mg
30453135 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455709 Supplier Toronto Research Chemicals Supplier No. TRCT369560500MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiosultap Sodium Salt
Molecular Formula C5H13NO6S4 • x(Na)
Quantity 500 mg
Synonym Bisultap, Dimehypo, Dimehypo Jumbo, Sodium 2-Dimethylaminopropane-1,3-bisthiosulfonate, Disosultap, Molosultap, Sha Chong Shuang, Thiosultapsodium, S,S'-(2-(Dimethylamino)propane-1,3-diyl) Bis(O-hydrogen sulfothioate) Sodium Salt
SMILES OS(SCC(N(C)C)CSS(O)(=O)=O)(=O)=O
Molecular Weight (g/mol) 311.422299
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.