missing translation for 'onlineSavingsMsg'
Learn More

Thiosalicylic acid, 98%

Product Code. p-200270 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10071590

Brand: Thermo Scientific Acros 138890050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 147-93-3 | C7H6O2S | 154.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-93-3
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.183
MDL Number MFCD00004836
InChI Key NBOMNTLFRHMDEZ-UHFFFAOYSA-N
Synonym thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto
PubChem CID 5443
ChEBI CHEBI:59124
IUPAC Name 2-sulfanylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)S

Specifications

Melting Point 164°C to 169°C
Color White to Yellow
Assay Percent Range 98%
Linear Formula HSC6H4CO2H
Beilstein 10, 125
Merck Index 15, 9513
Formula Weight 154.18
Percent Purity ≥97.5%
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Fine Crystalline Powder
Chemical Name or Material Thiosalicylic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.