missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioridazine hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Thioridazine hydrochloride is also a potent inhibitor of PI3K-Akt-mTOR signaling pathways with anti-angiogenic effect. Thioridazine hydrochloride shows antiproliferative and apoptosis induction effects in various types of cancer cells, with specificity on targeting cancer stem cells (CSCs).
Specifications
Specifications
| Chemical Name or Material | Thioridazine hydrochloride |
| Physical Form | Solid |
| CAS | 130-61-0 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H27ClN2S2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | H2O : 100 mg/mL (245.68 mM; Need ultrasonic) ∣DMSO : ≥ 45 mg/mL (110.55 mM) |
| SMILES | CSC(C=C1N2CCC3N(C)CCCC3)=CC=C1SC4=C2C=CC=C4.[H]Cl |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction