missing translation for 'onlineSavingsMsg'
Learn More

Thioridazine hydrochloride, MedChemExpress

Product Code. 16498536
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498536 100 mg 100mg
16492226 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498536 Supplier Medchem Express Supplier No. HYB0965
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Thioridazine hydrochloride, an orally active antagonist of the dopamine receptor D2 family proteins, exhibits potent anti-psychotic and anti-anxiety activities.

Thioridazine hydrochloride is also a potent inhibitor of PI3K-Akt-mTOR signaling pathways with anti-angiogenic effect. Thioridazine hydrochloride shows antiproliferative and apoptosis induction effects in various types of cancer cells, with specificity on targeting cancer stem cells (CSCs).

Specifications

Chemical Name or Material Thioridazine hydrochloride
Physical Form Solid
CAS 130-61-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H27ClN2S2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : 100 mg/mL (245.68 mM; Need ultrasonic) ∣DMSO : ≥ 45 mg/mL (110.55 mM)
SMILES CSC(C=C1N2CCC3N(C)CCCC3)=CC=C1SC4=C2C=CC=C4.[H]Cl
Molecular Weight (g/mol) 407.04
Formula Weight 407.04
Percent Purity 99.46%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.