missing translation for 'onlineSavingsMsg'
Learn More

Thioridazine.HCl, Lipomed

Product Code. 30459643
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30459643

Brand: Lipomed LPMTRD1839HC50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Thioridazine.HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thioridazine hydrochloride
CAS 130-61-0
Analyte or Component Names Thioridazine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H26 N2 S2 . Cl H
Synonym Thioridazine Hydrochloride
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CSc1ccc2Sc3ccccc3N(CCC4CCCCN4C)c2c1
IUPAC Name 10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfanylphenothiazine;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 407.04
Quantity 50 mg
Formula Weight 406.1304 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.