missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioredoxin Reductase 1/TRXR1 Mouse anti-Human, Clone: 30F1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59489-50UL
This item is not returnable.
View return policy
Description
Thioredoxin Reductase 1/TRXR1 Monoclonal specifically detects Thioredoxin Reductase 1/TRXR1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Thioredoxin Reductase 1/TRXR1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| EC 1.8.1, EC 1.8.1.9, Gene associated with retinoic and IFN-induced mortality 12 protein, Gene associated with retinoic and interferon-induced mortality 12 protein, GRIM12, GRIM-12TR1, KDRF, KM-102-derived reductase-like factor, MGC9145, oxidoreductase, thioredoxin reductase 1, thioredoxin reductase 1, cytoplasmic, thioredoxin reductase GRIM-12, Thioredoxin reductase TR1, TR, Trxr1, TXNRTRXR1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 30F1 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| TXNRD1 | |
| Recombinant human TXNRD1 (161-647aa) purified from E. coli | |
| 50 μL | |
| Breast Cancer, Lipid and Metabolism | |
| 7296 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction