missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioredoxin Reductase 1/TRXR1 Antibody (30F1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59489PE
This item is not returnable.
View return policy
Description
Thioredoxin Reductase 1/TRXR1 Monoclonal antibody specifically detects Thioredoxin Reductase 1/TRXR1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Thioredoxin Reductase 1/TRXR1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 30F1 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| EC 1.8.1, EC 1.8.1.9, Gene associated with retinoic and IFN-induced mortality 12 protein, Gene associated with retinoic and interferon-induced mortality 12 protein, GRIM12, GRIM-12TR1, KDRF, KM-102-derived reductase-like factor, MGC9145, oxidoreductase, thioredoxin reductase 1, thioredoxin reductase 1, cytoplasmic, thioredoxin reductase GRIM-12, Thioredoxin reductase TR1, TR, Trxr1, TXNRTRXR1 | |
| Recombinant human Thioredoxin Reductase 1/TRXR1 (161-647aa) purified from E. coli | |
| 0.1 mL | |
| Breast Cancer, Lipid and Metabolism | |
| 7296 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction