missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioredoxin-like 5/TRP14/TXNDC17 Rabbit anti-Human, Clone: 43, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89879-100UL
This item is not returnable.
View return policy
Description
Thioredoxin-like 5/TRP14/TXNDC17 Monoclonal specifically detects Thioredoxin-like 5/TRP14/TXNDC17 in Human samples. It is validated for ELISA.
Specifications
| Thioredoxin-like 5/TRP14/TXNDC17 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 84817 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 43 | |
| ELISA 1:5000-1:10000 | |
| MGC14353, Protein 42-9-9, thioredoxin (Trx)-related protein, 14 kDa, thioredoxin domain containing 17, thioredoxin domain-containing protein 17, thioredoxin-like 5, Thioredoxin-like protein 5, TRP1414 kDa thioredoxin-related protein, TXNL5 | |
| Recombinant Human Thioredoxin-like 5/TRP14/TXNDC17 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction