missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioredoxin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577048
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Thioredoxin 2 (Trx-2) is a small redox protein that is localized to the mitochondria and is essential for cell viability, playing a crucial role in the scavenging of ROS in mitochondria and regulating the mitochondrial apoptosis signaling pathway. Trx reductases (TrxR1 and TrxR2) are ubiquitously expressed flavoproteins that catalyze the NADPH-dependent reduction of Trx as well as several other oxidized cellular components. Mammalian Trx reductases are a part of a selenium-containing pyridine nucleotide-disulphide oxidoreductase family, which has a conserved catalytic site of Cys-Val-Asn-Val-Gly-Cys. TrxR1 and TrxR2 are also involved in the prevention of oxidative stress. Inhibition of TrxR activity may provide for potential treatments of cancer, AIDS and other autoimmune diseases as well as bacterial infections and parasitic diseases.
Specifications
| Thioredoxin 2 | |
| Polyclonal | |
| Unconjugated | |
| TXN2 | |
| Thioredoxin, mitochondrial; MTRX; Mt-Trx; Thioredoxin-2; Thioredoxin 2; Thioredoxin2; TXN2; TRX-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25828, 56551, 79462 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P97493, P97615, Q99757 | |
| TXN2 | |
| Recombit full length Human Thioredoxin 2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction