missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioredoxin-2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00013MFV500
This item is not returnable.
View return policy
Description
Thioredoxin-2 Polyclonal antibody specifically detects Thioredoxin-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| 50mM Sodium Borate | |
| Antigen and protein A Affinity-purified | |
| Store at 4C in the dark. |
| Rabbit | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction