missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioredoxin-1 Rabbit anti-Mouse, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90730-100UL
This item is not returnable.
View return policy
Description
Thioredoxin-1 Monoclonal specifically detects Thioredoxin-1 in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| Thioredoxin-1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Recombinant Mouse Thioredoxin-1 Protein | |
| 100 μg | |
| Breast Cancer, Lipid and Metabolism | |
| 7295 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 2 | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction