Learn More
Thioredoxin-1 Antibody (002), Janelia Fluor™ 549, Novus Biologicals™
Product Code. 18401749 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
Product Code. unitSize
18401749 0.1mL
1 options
Product Code. 18401749 Supplier Novus Biologicals Supplier No. NBP290730JF549
Rabbit Monoclonal Antibody
Thioredoxin-1 Monoclonal antibody specifically detects Thioredoxin-1 in Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Gene Alias TRDX;TRX;TRX1;Trx80;TXN
Gene ID (Entrez) 7295