missing translation for 'onlineSavingsMsg'
Learn More

Thioquinox, TRC

Product Code. 30457376
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457376 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457376 Supplier Toronto Research Chemicals Supplier No. TRCT368298500MG
 Restricted Item
Estimated Shipment: 19-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thioquinox
CAS 93-75-4
Molecular Formula C9 H4 N2 S3
Quantity 500 mg
Synonym 1,3-Dithiolo[4,5-b]quinoxaline-2-thione (9CI','ACI), Carbonic acid','trithio-','cyclic 2,3-quinoxalinediyl ester (8CI), Carbonic acid','trithio-','cyclic ester with 2,3-quinoxalinedithiol (6CI), 2,3-Quinoxalinedithiol cyclic trithiocarbonate, 2-Thioxo-1,3-dithiolo[4,5-b]quinoxaline, 2H-[1,3]Dithiolo[4,5-b]quinoxaline-2-thione, Bayer 30686, Chinothionat, Eradex, Quinothionate, Quinoxaline-2,3-diyl trithiocarbonate, Readex, Thiaquinox, Thioquinox
InChI Formula InChI=1S/C9H4N2S3/c12-9-13-7-8(14-9)11-6-4-2-1-3-5(6)10-7/h1-4H
SMILES S=C1Sc2nc3ccccc3nc2S1
IUPAC Name [1,3]dithiolo[4,5-b]quinoxaline-2-thione
Molecular Weight (g/mol) 236.34
Formula Weight 235.9537
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.