missing translation for 'onlineSavingsMsg'
Learn More

Thiophenol, 99+%, ACROS Organics™

Product Code. 10282982 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10282982 5mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10282982 Supplier Thermo Scientific Acros Supplier No. 220040050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 108-98-5
Density 1.0780g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C6H6S
Linear Formula C6H5SH
Refractive Index 1.5880 to 1.5900 (20°C, 589nm)
MDL Number MFCD00004826
Quantity 5 mL
Beilstein 06, 294
Fieser 06,585; 07,367; 09,465; 10,399; 12,485; 13,53; 16,327
Merck Index 15, 9508
Synonym thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto
InChI Key RMVRSNDYEFQCLF-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)S
IUPAC Name benzenethiol
Molecular Weight (g/mol) 110.174
PubChem CID 7969
ChEBI CHEBI:48498
Formula Weight 110.17
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material Thiophenol, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.