missing translation for 'onlineSavingsMsg'
Learn More

Thiophene, 99%

Product Code. 11481668
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481668 500 g 500g
11443930 50 g 50g
11453930 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481668 Supplier Thermo Scientific Alfa Aesar Supplier No. A13941.36
  • ONLINE EXCLUSIVE PRICE
    £58.70
    £52.65 / 500g
    Save £6.05
    10% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-02-1 | C4H4S | 84.14 g/mol

Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.

Solubility
Miscible with ethyl alcohol, diethyl ether and other organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-02-1
Molecular Formula C4H4S
Molecular Weight (g/mol) 84.14
MDL Number MFCD00005413
InChI Key YTPLMLYBLZKORZ-UHFFFAOYSA-N
Synonym thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
PubChem CID 8030
ChEBI CHEBI:30856
IUPAC Name thiophene
SMILES S1C=CC=C1

Specifications

Melting Point -38°C
Density 1.066
Boiling Point 83°C to 85°C
Flash Point −9°C (16°F)
Odor Stench
Refractive Index 1.528
Quantity 500 g
UN Number UN2414
Beilstein 103222
Merck Index 14,9353
Solubility Information Miscible with ethyl alcohol,diethyl ether and other organic solvents. Immiscible with water.
Formula Weight 84.14
Percent Purity 99%
Chemical Name or Material Thiophene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thiophene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Thiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.