missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-ethylamine, 98%

Product Code. 10467582 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10467582 1 g Glass bottle 1g
10633183 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10467582 Supplier Thermo Scientific Acros Supplier No. 354270010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30433-91-1 | C6H9NS | 127.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30433-91-1
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.21
MDL Number MFCD00051495
InChI Key HVLUYXIJZLDNIS-UHFFFAOYSA-N
Synonym thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene
PubChem CID 116521
IUPAC Name 2-thiophen-2-ylethanamine
SMILES C1=CSC(=C1)CCN

Specifications

Color Colorless to Yellow
Boiling Point 103.0°C to 105.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5490 to 1.5510
Quantity 1 g
Formula Weight 127.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Thiophene-2-ethylamine
Show More Show Less
compliance-icons
Product Identifier
  • Thiophene-2-ethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.