missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carboxylic anhydride, 95%

Product Code. p-7046026
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333406 1 g 1g
11343406 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333406 Supplier Thermo Scientific Alfa Aesar Supplier No. L12851.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25569-97-5 | C10H6O3S2 | 238.275 g/mol

Thiophene-2-carboxylic anhydride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carboxylic anhydride is used as an organic chemical synthesis intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25569-97-5
Molecular Formula C10H6O3S2
Molecular Weight (g/mol) 238.275
MDL Number MFCD00068089
InChI Key BKBCDDYQJQGMNX-UHFFFAOYSA-N
Synonym thiophene-2-carboxylic anhydride, thiophene-2-carboxylic acid anhydride, 2-thenoyl ether, 2-thenoic anhydride, acmc-20ajil, bis 2-thiophenecarboxylic acid anhydride, 2-thiophenecarboxylicacid,1,1'-anhydride, 2-thiophenecarboxylicacid, 1,1'-anhydride, thiophen-2-ylcarbonyl thiophene-2-carboxylate, 2-thiophenecarboxylic acid oxo thiophen-2-yl methyl ester
PubChem CID 3680768
IUPAC Name thiophene-2-carbonyl thiophene-2-carboxylate
SMILES C1=CSC(=C1)C(=O)OC(=O)C2=CC=CS2

Specifications

Melting Point 58°C to 61°C
Quantity 1 g
Beilstein 177049
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 238.29
Percent Purity 95%
Chemical Name or Material Thiophene-2-carboxylic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.