missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carboxylic acid, 99%

Product Code. 11464164
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464164 25 g 25g
11474164 100 g 100g
11484164 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464164 Supplier Thermo Scientific Alfa Aesar Supplier No. A12514.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 527-72-0 | C5H4O2S | 128.15 g/mol

A hypoglycemic, anti-lipolytic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-72-0
Molecular Formula C5H4O2S
Molecular Weight (g/mol) 128.15
MDL Number MFCD00005437
InChI Key QERYCTSHXKAMIS-UHFFFAOYSA-N
Synonym 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid
PubChem CID 10700
ChEBI CHEBI:71241
IUPAC Name thiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=CS1

Specifications

Melting Point 126°C to 130°C
Density 1.42
Boiling Point 259°C to 261°C
Quantity 25 g
Beilstein 110150
Merck Index 14,9354
Formula Weight 128.15
Percent Purity 99%
Chemical Name or Material Thiophene-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.