missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carboxaldehyde, 98+%

Product Code. 11413930
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
Unit Size:
1000g
100g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413930 100 g 100g
11423930 500 g 500g
11466907 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413930 Supplier Thermo Scientific Alfa Aesar Supplier No. A14344.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-03-3 | C5H4OS | 112.15 g/mol

Thiophene-2-carboxaldehyde is used in the preparation of beta-aryl-beta-amino acids and urea derivatives. It acts as an arylation reagent. Further, it is used in the synthesis of unsaturated ketones, which serve as antiviral and cytotoxic agents. In addition to this, it is used as an intermediate to manufacture pharmaceuticals and aromatic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carboxaldehyde is used in the preparation of beta-aryl-beta-amino acids and urea derivatives. It acts as an arylation reagent. Further, it is used in the synthesis of unsaturated ketones, which serve as antiviral and cytotoxic agents. In addition to this, it is used as an intermediate to manufacture pharmaceuticals and aromatic compounds.

Solubility
Miscible with chloroform, alcohol, ether, dimethylsulfoxide, ethyl acetate and most organic solvents. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-03-3
Molecular Formula C5H4OS
Molecular Weight (g/mol) 112.15
MDL Number MFCD00005429
InChI Key CNUDBTRUORMMPA-UHFFFAOYSA-N
Synonym 2-thiophenecarboxaldehyde, 2-thenaldehyde, thiophene-2-aldehyde, 2-formylthiophene, 2-thienylaldehyde, 2-thiophenealdehyde, thiophene-2-carboxaldehyde, 2-thienylcarboxaldehyde, 2-thienaldehyde, alpha-formylthiophene
PubChem CID 7364
ChEBI CHEBI:87301
IUPAC Name thiophene-2-carbaldehyde
SMILES O=CC1=CC=CS1

Specifications

Density 1.22
Boiling Point 196°C to 198°C
Flash Point 78°C (172°F)
Refractive Index 1.591
Quantity 100 g
Beilstein 105819
Sensitivity Air sensitive
Solubility Information Miscible with chloroform,alcohol,ether,dimethylsulfoxide,ethyl acetate and most organic solvents. Immiscible with water.
Formula Weight 112.15
Percent Purity ≥98%
Chemical Name or Material Thiophene-2-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • Thiophene-2-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Thiophenecarboxaldehyde

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.