missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carbonyl chloride, 98%

Product Code. 11427596
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427596 25 g 25g
11437596 100 g 100g
11447596 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427596 Supplier Thermo Scientific Alfa Aesar Supplier No. A14641.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5271-67-0 | C5H3ClOS | 146.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5271-67-0
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.59
MDL Number MFCD00005428
InChI Key QIQITDHWZYEEPA-UHFFFAOYSA-N
Synonym 2-thiophenecarbonyl chloride, 2-thenoyl chloride, 2-thienylcarbonyl chloride, 2-thiophenecarbonylchloride, thiophenecarbonyl chloride, 2-thiophene carbonyl chloride, thiophene-2-carboxylic acid chloride, thenoyl chloride, thienoyl chloride, 2-thenoylchloride
PubChem CID 78928
IUPAC Name thiophene-2-carbonyl chloride
SMILES ClC(=O)C1=CC=CS1

Specifications

Density 1.372
Boiling Point 105°C (30 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.59
Quantity 25 g
UN Number UN3265
Beilstein 110145
Sensitivity Moisture sensitive
Formula Weight 146.6
Percent Purity 98%
Chemical Name or Material Thiophene-2-carbonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.