missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carbonitrile, 98%, Thermo Scientific Chemicals

Catalog Number 11473914
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
1003-31-2
C5H3NS
109.146
MFCD00005416
CUPOOAWTRIURFT-UHFFFAOYSA-N
2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile
66087
thiophene-2-carbonitrile
C1=CSC(=C1)C#N
This item is not returnable. View return policy
1003-31-2
C5H3NS
109.146
MFCD00005416
CUPOOAWTRIURFT-UHFFFAOYSA-N
2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile
66087
thiophene-2-carbonitrile
C1=CSC(=C1)C#N

Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2'-thienylpyrroles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2′-thienylpyrroles.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Density 1.171
Boiling Point 191°C to 193°C
Flash Point 81°C (178°F)
Odor Unpleasant
Refractive Index 1.5645
Quantity 500 g
UN Number UN3276
Beilstein 107035
Solubility Information Slightly miscible with water.
Formula Weight 109.15
Percent Purity 98%
Chemical Name or Material Thiophene-2-carbonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'