missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10341995
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10341995 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10341995 Supplier Thermo Scientific Maybridge Supplier No. CC13004EB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 98-03-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H4OS
MDL Number MFCD00005429
Quantity 25 g
Synonym 2-thiophenecarboxaldehyde, 2-thenaldehyde, thiophene-2-aldehyde, 2-formylthiophene, 2-thienylaldehyde, 2-thiophenealdehyde, thiophene-2-carboxaldehyde, 2-thienylcarboxaldehyde, 2-thienaldehyde, alpha-formylthiophene
InChI Key CNUDBTRUORMMPA-UHFFFAOYSA-N
SMILES O=CC1=CC=CS1
IUPAC Name thiophene-2-carbaldehyde
Molecular Weight (g/mol) 112.15
PubChem CID 7364
ChEBI CHEBI:87301
Formula Weight 112.15
Percent Purity 97%
Chemical Name or Material Thiophene-2-carbaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.