missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2,5-diboronic acid bis(pinacol) ester, 97%

Product Code. 15449295
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449295 5 g 5g
15439295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449295 Supplier Thermo Scientific Alfa Aesar Supplier No. H51725.06
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 175361-81-6 | C16H26B2O4S | 336.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175361-81-6
Molecular Formula C16H26B2O4S
Molecular Weight (g/mol) 336.06
MDL Number MFCD09800563
InChI Key AOJXAKMKFDBHHT-UHFFFAOYSA-N
Synonym 2,5-bis-thiopheneboronic acid pinacol ester, 2,5-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene, thiophene-2,5-diboronic acid bis pinacol ester, thiophene-2,5-diboronicacidbis pinacol ester, bis 4,4,5,5-tetramethyl-1,3-dioxaborolane-2,5-thiophene, 2,5-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl thiophene, 2,2'-thiene-2,5-diyl bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl-1,3,2-dioxaborolane
PubChem CID 23058061
IUPAC Name 4,4,5,5-tetramethyl-2-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(S1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 216°C to 220°C
Quantity 5 g
Formula Weight 336.06
Percent Purity 97%
Chemical Name or Material Thiophene-2,5-diboronic acid bis(pinacol) ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.