missing translation for 'onlineSavingsMsg'
Learn More

Thiophanate-methyl, Dr. Ehrenstorfer

Product Code. 30321726
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30321726

Brand: Dr. Ehrenstorfer DREC17545000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiophanate-methyl
CAS 23564-05-8
Analyte or Component Names Thiophanate-methyl
Shipping Condition +20°C
Molecular Formula C12 H14 N4 O4 S2
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C12H14N4O4S2/c1-19-11(17)15-9(21)13-7-5-3-4-6-8(7)14-10(22)16-12(18)20-2/h3-6H,1-2H3,(H2,13,15,17,21)(H2,14,16,18,22)
SMILES COC(=O)NC(=S)Nc1ccccc1NC(=S)NC(=O)OC
IUPAC Name methyl N-[[2-(methoxycarbonylcarbamothioylamino)phenyl]carbamothioyl]carbamate
Molecular Weight (g/mol) 342.39
Quantity 100 mg
Formula Weight 342.0456 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.