missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Thioperamide

Description
Potent histamine H3 and H4 antagonist/inverse agonist. Ki values are 25 and 27 nM for human recombinant H3 and H4 receptors respectively. Blocks eosinophil shape change (IC50 = 1.4 μM) and chemotaxis (IC50 = 519 nM) induced by histamine. Freely crosses the blood-brain barrier.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 292.44 |
| Purity | >99% |
| Molecular Weight (g/mol) | 408.517 |
| Chemical Name or Material | Thioperamide |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?